3-Fluor-2-methoxybenzoesäure

Lieferant: FLUOROCHEM

CFF1123. 013029-1G 013029-5G
CHMPCFF1123.EA 0 EUR
CHMPCFF1123. FLUO013029-1G FLUO013029-5G
3-Fluor-2-methoxybenzoesäure
3-Fluoro-2-methoxybenzoic acid
Formel: C₈H₇FO₃
Molecular Weight: 170.14 g/mol
MDL: MFCD00973972
CAS-Nummer: 106428-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR