2,4,5-Trifluor-3-methoxybenzoesäure

Lieferant: FLUOROCHEM

Synonyme: Moxidectin Impurity 5, 3-Methoxy-2,4,5-trifluorobenzoic acid

009272-10G 009272-50G
FLUO009272-10GEA 0 EUR
FLUO009272-10G FLUO009272-50G
2,4,5-Trifluor-3-methoxybenzoesäure
2,4,5-Trifluor-3-methoxybenzoesäure
Formel: C₈H₅F₃O₃
Molecular Weight: 206.12 g/mol
MDL: MFCD00153201
CAS-Nummer: 112811-65-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR