2,3-Difluor-4-hydroxybenzoesäure

Lieferant: FLUOROCHEM

013021-1G 013021-2.5G
FLUO013021-1GEA 125 EUR
FLUO013021-1G FLUO013021-2.5G
2,3-Difluor-4-hydroxybenzoesäure
2,3-Difluoro-4-hydroxybenzoic acid
Formel: C₇H₄F₂O₃
MDL: MFCD01321116
CAS-Nummer: 175968-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR