3-Fluorsalicylsäure

Lieferant: FLUOROCHEM

013028-1G 013028-5G
FLUO013028-1GEA 57.9 EUR
FLUO013028-1G FLUO013028-5G
3-Fluorsalicylsäure
3-Fluorsalicylsäure
Formel: C₇H₅FO₃
Molecular Weight: 156,11 g/mol
MDL: MFCD00153167
CAS-Nummer: 341-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR