3-Fluor-4-hydroxybenzoesäure

Lieferant: FLUOROCHEM

013030-1G 013030-5G
FLUO013030-1GEA 31.1 EUR
FLUO013030-1G FLUO013030-5G
3-Fluor-4-hydroxybenzoesäure
3-Fluoro-4-hydroxybenzoic acid
Formel: C₇H₅FO₃
Molecular Weight: 156,11 g/mol
MDL: MFCD00143161
CAS-Nummer: 350-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR