3-Fluor-4-hydroxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

544809-1G 544809-5G
SIAL544809-1GEA 53.6 EUR
SIAL544809-1G SIAL544809-5G
3-Fluor-4-hydroxybenzoesäure, Sigma-Aldrich®
3-Fluoro-4-hydroxybenzoic acid
Formel: C₇H₅FO₃
Molecular Weight: 156,11 g/mol
MDL: MFCD00143161
CAS-Nummer: 350-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR