2,3,5,6-Tetrafluor-4-hydroxybenzoesäure
Lieferant: FLUOROCHEM
008803-1G
008803-5G
008803-25G
FLUO008803-1GEA
0
EUR
FLUO008803-1G
FLUO008803-5G
FLUO008803-25G
2,3,5,6-Tetrafluor-4-hydroxybenzoesäure
2,3,5,6-Tetrafluor-4-hydroxybenzoesäure
Formel:
C₇H₂F₄O₃ Molecular Weight: 210.08 g/mol |
MDL:
MFCD00129952 CAS-Nummer: 652-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...