2-((Difluormethyl)thio)benzoesäure
Lieferant: CHEMPUR
FL5768.5
FL5768.10
005768-1G
005768-5G
CHMPFL5768.5EA
0
EUR
CHMPFL5768.5
CHMPFL5768.10
FLUO005768-1G
FLUO005768-5G
2-((Difluormethyl)thio)benzoesäure
2-((Difluormethyl)thio)benzoesäure
Formel:
C₈H₆F₂O₂S Molecular Weight: 204,2 g/mol Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 149…150 °C |
CAS-Nummer:
79676-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...