4-((Trifluormethyl)thio)benzoesäure

Lieferant: FLUOROCHEM

007999-10G 007999-5G
FLUO007999-10GEA 0 EUR
FLUO007999-10G FLUO007999-5G
4-((Trifluormethyl)thio)benzoesäure
4-((Trifluormethyl)thio)benzoesäure
Formel: C₈H₅F₃O₂S
MDL: MFCD00040906
CAS-Nummer: 330-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR