2-Fluor-5-iodbenzoesäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H26175.03
H26175.06
H26175.03EA
29.8
EUR
H26175.03
H26175.06
2-Fluor-5-iodbenzoesäure ≥97%
2-Fluor-5-iodbenzoesäure
Purity: 97%
CAS No.: 124700-41-0
Molecular Formula: C7H4FIO2
Formula Weight: 266.01
Melting Point: 162-165°
MDL No.: MFCD03094517
Formel:
C₇H₄FIO₂ Schmelzpunkt: 162…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094517 CAS-Nummer: 124700-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...