2-Fluor-5-iodbenzoesäure

Lieferant: FLUOROCHEM
Achtung

013023-5G 013023-25G 013023-1G
FLUO013023-5GEA 53.9 EUR
FLUO013023-5G FLUO013023-25G FLUO013023-1G
2-Fluor-5-iodbenzoesäure
2-Fluor-5-iodbenzoesäure
Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
MDL: MFCD03094517
CAS-Nummer: 124700-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR