1-Iod-2-(trifluormethoxy)benzol
Lieferant: Thermo Scientific
TL00358.1
TL00358.10
TL00358EE
MAYBTL00358.1EA
0
EUR
MAYBTL00358.1
MAYBTL00358.10
MAYBTL00358EE
1-Iod-2-(trifluormethoxy)benzol
1-Iod-2-(trifluormethoxy)benzol
Formel:
C₇H₄F₃IO Molecular Weight: 288,01 g/mol |
MDL:
MFCD00042410 CAS-Nummer: 175278-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...