1-Iod-4-(trifluormethoxy)benzol
Lieferant: Thermo Scientific
Synonyme:
4-(Trifluormethoxy)iodbenzol
TL00338.1
TL00338.10
TL00338.50
MAYBTL00338.1EA
0
EUR
MAYBTL00338.1
MAYBTL00338.10
MAYBTL00338.50
1-Iod-4-(trifluormethoxy)benzol
1-Iod-4-(trifluormethoxy)benzol
Formel:
C₇H₄F₃IO Molecular Weight: 288,01 g/mol |
MDL:
MFCD00042411 CAS-Nummer: 103962-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...