1-Iod-4-(trifluormethoxy)benzol

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluormethoxy)iodbenzol

007558-1G 007558-25G 007558-100G
FLUO007558-1GEA 31.1 EUR
FLUO007558-1G FLUO007558-25G FLUO007558-100G
1-Iod-4-(trifluormethoxy)benzol
1-Iod-4-(trifluormethoxy)benzol
Formel: C₇H₄F₃IO
Molecular Weight: 288.01 g/mol
MDL: MFCD00042411
CAS-Nummer: 103962-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR