2,3,4-Trifluorphenylisocyanat

Lieferant: Thermo Scientific

AC31950.250 AC31950.1
MAYBAC31950.250EA 0 EUR
MAYBAC31950.250 MAYBAC31950.1
2,3,4-Trifluorphenylisocyanat
2,3,4-Trifluorphenylisocyanat
Formel: C₇H₂F₃NO
Molecular Weight: 173,09 g/mol
CAS-Nummer: 190774-58-4
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR