2-(Trifluormethoxy)phenylisocyanat

Lieferant: Thermo Scientific

Synonyme: 2-Trifluoromethoxyphenylisocyanate, Isocyanic acid 2-(Trifluoromethoxy)phenyl ester

SB01819.1 SB01819.10
MAYBSB01819.1EA 0 EUR
MAYBSB01819.1 MAYBSB01819.10
2-(Trifluormethoxy)phenylisocyanat
2-(Trifluormethoxy)phenylisocyanat
Formel: C₈H₄F₃NO₂
Molecular Weight: 203.12 g/mol
Siedepunkt: 52 °C (15 mmHg)
Dichte: 1.35
Lagertemperatur: Kühlschrank
MDL: MFCD00134160
CAS-Nummer: 182500-26-1
UN: 3080
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR