2,6-Difluorphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1,3-Difluor-2-isothiocyanatobenzol
005374-1G
005374-5G
005374-25G
FLUO005374-1GEA
45.9
EUR
FLUO005374-1G
FLUO005374-5G
FLUO005374-25G
2,6-Difluorphenylisothiocyanat
2,6-Difluorphenylisothiocyanat
Formel:
C₇H₃F₂NS Molecular Weight: 171,17 g/mol Siedepunkt: 106…107 °C (35 mmHg) Lagertemperatur: Kühlschrank |
MDL:
MFCD00041047 CAS-Nummer: 207974-17-2 UN: 2930 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...