2,4-Difluorphenylisothiocyanat

Lieferant: CHEMPUR
Achtung

Synonyme: 1,3-Difluor-4-isothiocyanatobenzol

FL8856.POR 008856-5G 008856-50G
CHMPFL8856.POREA 0 EUR
CHMPFL8856.POR FLUO008856-5G FLUO008856-50G
2,4-Difluorphenylisothiocyanat
2,4-Difluorphenylisothiocyanat
Formel: C₇H₃F₂NS
Molecular Weight: 171,17 g/mol
Siedepunkt: 98…99 °C (30 mmHg)
Dichte: 1,349 g/cm³ (20 °C)
MDL: MFCD00041044
CAS-Nummer: 141106-52-7
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR