3,5-Difluorphenylisothiocyanat

Lieferant: FLUOROCHEM

009609-1G 009609-5G
FLUO009609-1GEA 55.4 EUR
FLUO009609-1G FLUO009609-5G
3,5-Difluorphenylisothiocyanat
3,5-Difluorphenylisothiocyanat
Formel: C₇H₃F₂NS
Molecular Weight: 171,17 g/mol
CAS-Nummer: 302912-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR