2,5-Difluorphenylisothiocyanat

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 1,4-Difluoro-2-isothiocyanatobenzene

008857-5G 008857-25G
FLUO008857-5GEA 219 EUR
FLUO008857-5G FLUO008857-25G
2,5-Difluorphenylisothiocyanat
2,5-Difluorphenylisothiocyanat
Formel: C₇H₃F₂NS
Molecular Weight: 171,17 g/mol
Siedepunkt: 111…112 °C (35 mmHg)
Dichte: 1,338 g/cm³ (20 °C)
MDL: MFCD00041046
CAS-Nummer: 206559-57-1
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR