2,4,6-Trifluorphenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1-Isothiocyanato-2,4,6-trifluorbenzol

009581-1G 009581-5G
FLUO009581-1GEA 63.9 EUR
FLUO009581-1G FLUO009581-5G
2,4,6-Trifluorphenylisothiocyanat
2,4,6-Trifluorphenylisothiocyanat
Formel: C₇H₂F₃NS
Molecular Weight: 189,16 g/mol
Siedepunkt: 111…112 °C (35 mmHg)
MDL: MFCD00041045
CAS-Nummer: 206761-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR