3,4,5-Trifluorphenylisothiocyanat

Lieferant: FLUOROCHEM

009600-1G 009600-5G
FLUO009600-1GEA 84.9 EUR
FLUO009600-1G FLUO009600-5G
3,4,5-Trifluorphenylisothiocyanat
3,4,5-Trifluorphenylisothiocyanat
Formel: C₇H₂F₃NS
CAS-Nummer: 362690-52-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR