4-(Trifluormethoxy)phenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluoromethoxy)phenylisothiocyanate
007569-1G
007569-2.5G
FLUO007569-1GEA
34.1
EUR
FLUO007569-1G
FLUO007569-2.5G
4-(Trifluormethoxy)phenylisothiocyanat
4-(Trifluormethoxy)phenylisothiocyanat
Formel:
C₈H₄F₃NOS Molecular Weight: 219,19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00042412 CAS-Nummer: 64285-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...