2,3-Difluor-6-nitroanilin ≥98%

Lieferant: FLUOROCHEM

Synonyme: 1-Amino-2,3-difluoro-6-nitrobenzene

008168-1G 008168-5G
FLUO008168-1GEA 31.2 EUR
FLUO008168-1G FLUO008168-5G
2,3-Difluor-6-nitroanilin ≥98%
2,3-Difluor-6-nitroanilin
Formel: C₆H₄F₂N₂O₂
MDL: MFCD00233313
CAS-Nummer: 211693-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR