2-Fluor-4-nitroanilin

Lieferant: CHEMPUR
Achtung

FL13094.POR 013094-25G 013094-5G 013094-10G 013094-1G
CHMPFL13094.POREA 0 EUR
CHMPFL13094.POR FLUO013094-25G FLUO013094-5G FLUO013094-10G FLUO013094-1G
2-Fluor-4-nitroanilin
2-Fluor-4-nitroanilin
Formel: C₆H₅FN₂O₂
Molecular Weight: 156,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00034560
CAS-Nummer: 369-35-7
EINECS: 206-719-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR