3-(Trifluormethoxy)anilin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Trifluoromethoxyaniline

A19592.03 A19592.06
A19592.03EA 43 EUR
A19592.03 A19592.06
3-(Trifluormethoxy)anilin ≥98%
3-(Trifluormethoxy)anilin

Purity: 98% CAS No.: 1535-73-5 Molecular Formula: C7H6F3NO Formula Weight: 177.13 Flash Point: 85°(185°F) Refractive Index: 1.4660 Odour: Characteristic Boiling Point: 72-75°/8mm Beilstein Reference: 776921 UN No.: 2810 MDL No.: MFCD00041511

Formel: C₇H₆F₃NO
Molecular Weight: 177,13 g/mol
Siedepunkt: 72…75 °C (8 mmHg)
Dichte: 1,325
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041511
CAS-Nummer: 1535-73-5
EINECS: 216-256-3
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR