4-(Difluormethoxy)anilin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
B20349.09
B20349.88
B20349.09EA
192
EUR
B20349.09
B20349.88
4-(Difluormethoxy)anilin ≥97%
4-(Difluormethoxy)anilin
Purity: 97%
CAS No.: 22236-10-8
Molecular Formula: C7H7F2NO
Formula Weight: 159.14
Flash Point:
Formel:
C₇H₇F₂NO Molecular Weight: 159,14 g/mol Siedepunkt: 231 °C Dichte: 1,286 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085005 CAS-Nummer: 22236-10-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...