4-(Trifluormethoxy)anilin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A17208.06
A17208.14
A17208.06EA
38.7
EUR
A17208.06
A17208.14
4-(Trifluormethoxy)anilin ≥98%
4-(Trifluormethoxy)anilin
Purity: 98%
CAS No.: 461-82-5
Molecular Formula: C7H6F3NO
Formula Weight: 177.13
Flash Point: 81°(178°F)
Refractive Index: 1.4630
Odour: Characteristic
Boiling Point: 73-75°/10mm
Beilstein Reference: 2090209
UN No.: 2810
MDL No.: MFCD00041314
Formel:
C₇H₆F₃NO Molecular Weight: 177,13 g/mol Siedepunkt: 73…75 °C (10 mmHg) Dichte: 1,310 Flammpunkt: 81 °C (178 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041314 CAS-Nummer: 461-82-5 EINECS: 207-317-5 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...