4-Amino-3-fluorphenol 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Fluoro-4-hydroxyaniline

H26084.MD H26084.03
H26084.MDEA 66.2 EUR
H26084.MD H26084.03
4-Amino-3-fluorphenol 96%
4-Amino-3-fluorphenol

Purity: 96% CAS No.: 399-95-1 Molecular Formula: C6H6FNO Formula Weight: 127.12 Melting Point: 137-140° UN No.: 3077 MDL No.: MFCD00077452

Formel: C₆H₆FNO
Molecular Weight: 127.12 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077452
CAS-Nummer: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR