4-Amino-3-fluorphenol

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 2-Fluoro-4-hydroxyaniline

CFF313.2.5 CFF313.5 003988-1G 003988-5G 003988-500MG
CHMPCFF313.2.5EA 427 EUR
CHMPCFF313.2.5 CHMPCFF313.5 FLUO003988-1G FLUO003988-5G FLUO003988-500MG
4-Amino-3-fluorphenol
4-Amino-3-fluorphenol
Formel: C₆H₆FNO
Molecular Weight: 127.12 g/mol
MDL: MFCD00077452
CAS-Nummer: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR