4-Amino-2-fluorphenol

Lieferant: FLUOROCHEM
Achtung

013049-1G 013049-5G 013049-500MG
FLUO013049-1GEA 31.1 EUR
FLUO013049-1G FLUO013049-5G FLUO013049-500MG
4-Amino-2-fluorphenol
4-Amino-2-fluorphenol
Formel: C₆H₆FNO
Molecular Weight: 127.12 g/mol
MDL: MFCD00671760
CAS-Nummer: 399-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR