2-Amino-4-fluorphenol 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Fluor-2-hydroxyanilin

H26165.03 H26165.06
H26165.03EA 49 EUR
H26165.03 H26165.06
2-Amino-4-fluorphenol 97%
2-Amino-4-fluorphenol

Purity: 97% CAS No.: 399-97-3 Molecular Formula: C6H6FNO Formula Weight: 127.12 Melting Point: 138-141° MDL No.: MFCD00077451

Formel: C₆H₆FNO
Schmelzpunkt: 138…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077451
CAS-Nummer: 399-97-3
EINECS: None

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR