2-Amino-4-fluorphenol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 5-Fluor-2-hydroxyanilin

013048-1G 013048-5G
FLUO013048-1GEA 31.1 EUR
FLUO013048-1G FLUO013048-5G
2-Amino-4-fluorphenol
2-Amino-4-fluorphenol
Formel: C₆H₆FNO
MDL: MFCD00077451
CAS-Nummer: 399-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR