1,3-Difluor-5-nitrobenzol

Lieferant: FLUOROCHEM
Achtung

011912-5G 011912-25G
FLUO011912-5GEA 0 EUR
FLUO011912-5G FLUO011912-25G
1,3-Difluor-5-nitrobenzol
1,3-Difluor-5-nitrobenzol
Formel: C₆H₃F₂NO₂
Molecular Weight: 159,09 g/mol
MDL: MFCD00012142
CAS-Nummer: 2265-94-3
EINECS: 218-867-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR