1-(Difluormethoxy)-4-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 4-(Difluoromethoxy)nitrobenzene

004840-5G 004840-100G 004840-25G
FLUO004840-5GEA 76.2 EUR
FLUO004840-5G FLUO004840-100G FLUO004840-25G
1-(Difluormethoxy)-4-nitrobenzol
1-(Difluormethoxy)-4-nitrobenzol
Formel: C₇H₅F₂NO₃
Molecular Weight: 189,12 g/mol
MDL: MFCD00085009
CAS-Nummer: 1544-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR