1-Nitro-3-(trifluormethoxy)benzol

Lieferant: Thermo Scientific

TL00391.1 TL00391.10
MAYBTL00391.1EA 0 EUR
MAYBTL00391.1 MAYBTL00391.10
1-Nitro-3-(trifluormethoxy)benzol
1-Nitro-3-(trifluormethoxy)benzol
Formel: C₇H₄F₃NO₃
Molecular Weight: 207.11 g/mol
MDL: MFCD00041011
CAS-Nummer: 2995-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR