1-Nitro-3-(trifluormethoxy)benzol
Lieferant: FLUOROCHEM
007560-1G
007560-5G
FLUO007560-1GEA
0
EUR
FLUO007560-1G
FLUO007560-5G
1-Nitro-3-(trifluormethoxy)benzol
1-Nitro-3-(trifluormethoxy)benzol
Formel:
C₇H₄F₃NO₃ Molecular Weight: 207.11 g/mol |
MDL:
MFCD00041011 CAS-Nummer: 2995-45-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...