1-Nitro-4-(trifluormethoxy)benzol
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluoromethoxy)nitrobenzene
007561-1G
007561-5G
007561-25G
FLUO007561-1GEA
31.1
EUR
FLUO007561-1G
FLUO007561-5G
FLUO007561-25G
1-Nitro-4-(trifluormethoxy)benzol
1-Nitro-4-(trifluormethoxy)benzol
Formel:
C₇H₄F₃NO₃ |
MDL:
MFCD00040302 CAS-Nummer: 713-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...