3-Fluorphenetol
Lieferant: CHEMPUR
Synonyme:
1-Fluor-3-ethoxybenzol, (3-Fluorphenyl)ethylether
FL13375.POR
013375-1G
013375-5G
CHMPFL13375.POREA
0
EUR
CHMPFL13375.POR
FLUO013375-1G
FLUO013375-5G
3-Fluorphenetol
3-Fluorphenetol
Formel:
C₈H₉FO Molecular Weight: 140,16 g/mol |
MDL:
MFCD00673011 CAS-Nummer: 458-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...