2-(Trifluormethoxy)phenol

Lieferant: FLUOROCHEM

007981-5G 007981-25G
FLUO007981-5GEA 34.1 EUR
FLUO007981-5G FLUO007981-25G
2-(Trifluormethoxy)phenol
2-(Trifluormethoxy)phenol
Formel: C₇H₅F₃O₂
MDL: MFCD00236324
CAS-Nummer: 32858-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR