3-(Trifluormethoxy)phenol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
α,α,α-Trifluor-3-hydroxyanisol
B20196.03
B20196.06
B20196.03EA
29.3
EUR
B20196.03
B20196.06
3-(Trifluormethoxy)phenol ≥98%
3-(Trifluormethoxy)phenol
Purity: 98%
CAS No.: 827-99-6
Molecular Formula: C7H5F3O2
Formula Weight: 178.11
Flash Point: 84°(183°F)
Refractive Index: 1.4460
Boiling Point: 68-69°/12mm
Beilstein Reference: 1868036
MDL No.: MFCD00040987
Formel:
C₇H₅F₃O₂ Molecular Weight: 178,11 g/mol Siedepunkt: 68…69 °C (12 mmHg) Dichte: 1,379 Flammpunkt: 84 °C (183 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040987 CAS-Nummer: 827-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...