3-(Trifluormethoxy)phenol

Lieferant: FLUOROCHEM

Synonyme: α,α,α-Trifluor-3-hydroxyanisol

002129-1G 002129-5G 002129-25G
FLUO002129-1GEA 31.1 EUR
FLUO002129-1G FLUO002129-5G FLUO002129-25G
3-(Trifluormethoxy)phenol
3-(Trifluormethoxy)phenol
Formel: C₇H₅F₃O₂
Molecular Weight: 178.11 g/mol
Siedepunkt: 69.5…79.5 °C (24 mmHg)
Dichte: 1.37 g/cm³ (20 °C)
MDL: MFCD00040987
CAS-Nummer: 827-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR