2-(Trifluormethoxy)thiophenol

Lieferant: CHEMPUR

Synonyme: 2-(Trifluoromethoxy)benzene-1-thiol, 2-(Trifluoromethoxy)benzenethiol

TL00363.1 TL00363.10
CHMPTL00363.1EA 0 EUR
CHMPTL00363.1 CHMPTL00363.10
2-(Trifluormethoxy)thiophenol
2-(Trifluormethoxy)thiophenol
Formel: C₇H₅F₃OS
Molecular Weight: 194.18 g/mol
MDL: MFCD00052320
CAS-Nummer: 175278-01-0
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR