4-(Trifluormethoxy)thiophenol

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluoromethoxy)benzenethiol, 4-(Trifluoromethoxy)benzene-1-thiol

007575-1G 007575-5G
FLUO007575-1GEA 94.1 EUR
FLUO007575-1G FLUO007575-5G
4-(Trifluormethoxy)thiophenol
4-(Trifluormethoxy)thiophenol
Formel: C₇H₅F₃OS
Molecular Weight: 194.18 g/mol
MDL: MFCD00052312
CAS-Nummer: 169685-29-4
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR