2-(Trifluormethoxy)benzolsulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 2-(Trifluormethoxy)benzolsulfochlorid, 2-(Trifluormethoxy)benzol-1-sulfonylchlorid, 2-(Trifluormethoxy)benzolsulfonsäurechlorid

TL00357.0001 TL00357.250 TL00357DA
MAYBTL00357.0001EA 0 EUR
MAYBTL00357.0001 MAYBTL00357.250 MAYBTL00357DA
2-(Trifluormethoxy)benzolsulfonylchlorid
2-(Trifluormethoxy)benzolsulfonylchlorid
Formel: C₇H₄ClF₃O₃S
Molecular Weight: 260,62 g/mol
MDL: MFCD00052316
CAS-Nummer: 103008-51-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR