2-(Trifluormethoxy)benzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluormethoxy)benzolsulfochlorid, 2-(Trifluormethoxy)benzol-1-sulfonylchlorid, 2-(Trifluormethoxy)benzolsulfonsäurechlorid

007539-1G 007539-5G 007539-25G
FLUO007539-1GEA 31.1 EUR
FLUO007539-1G FLUO007539-5G FLUO007539-25G
2-(Trifluormethoxy)benzolsulfonylchlorid
2-(Trifluormethoxy)benzolsulfonylchlorid
Formel: C₇H₄ClF₃O₃S
MDL: MFCD00052316
CAS-Nummer: 103008-51-1
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR