2,4-Difluorthiophenol

Lieferant: FLUOROCHEM

Synonyme: 1,3-Difluor-4-mercaptobenzol, 2,4-Difluorbenzolthiol, 2,4-Difluorbenzol-1-thiol, 2,4-Difluorphenylmercaptan

004878-1G 004878-100G 004878-25G
FLUO004878-1GEA 31.1 EUR
FLUO004878-1G FLUO004878-100G FLUO004878-25G
2,4-Difluorthiophenol
2,4-Difluorthiophenol
Formel: C₆H₄F₂S
Molecular Weight: 146,16 g/mol
Siedepunkt: 59 °C (1013 hPa)
Dichte: 1,29 g/cm³ (20 °C)
MDL: MFCD00203477
CAS-Nummer: 1996-44-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR