3-Chlor-4-fluorthiophenol
Lieferant: FLUOROCHEM
Synonyme:
1-Chlor-2-fluor-5-mercaptobenzol, 3-Chlor-4-fluorphenylmercaptan, 3-Chlor-4-fluorbenzol-1-thiol, 3-Chlor-4-fluorbenzolthiol
004469-1G
004469-5G
FLUO004469-1GEA
97.2
EUR
FLUO004469-1G
FLUO004469-5G
3-Chlor-4-fluorthiophenol
3-Chlor-4-fluorthiophenol
Formel:
C₆H₄ClFS |
MDL:
MFCD00052938 CAS-Nummer: 60811-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...