2-Chlor-4-fluorthiophenol

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-4-fluorbenzolthiol, 1-Chlor-3-fluor-6-mercaptobenzol, 2-Chlor-4-fluorphenylmercaptan, 2-Chlor-4-fluorbenzol-1-thiol

008695-1G 008695-5G
FLUO008695-1GEA 55.4 EUR
FLUO008695-1G FLUO008695-5G
2-Chlor-4-fluorthiophenol
2-Chlor-4-fluorthiophenol
Formel: C₆H₄ClFS
Molecular Weight: 162,61 g/mol
Siedepunkt: 86…88 °C (15 mmHg)
MDL: MFCD00052936
CAS-Nummer: 175277-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR