2,3,5,6-Tetrafluorthiophenol

Lieferant: Thermo Scientific

Synonyme: 2,3,5,6-Tetrafluorbenzolthiol, 2,3,5,6-Tetrafluorphenylmercaptan, 1-Mercapto-2,3,5,6-tetrafluorbenzol, 2,3,5,6-Tetrafluorbenzol-1-thiol

KM07934.1 KM07934.10
MAYBKM07934.1EA 0 EUR
MAYBKM07934.1 MAYBKM07934.10
2,3,5,6-Tetrafluorthiophenol
2,3,5,6-Tetrafluorthiophenol
Formel: C₆H₂F₄S
Molecular Weight: 182,14 g/mol
Siedepunkt: 152 °C (1013 hPa)
MDL: MFCD00004829
CAS-Nummer: 769-40-4
EINECS: 212-210-1
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR